Anti-TYRP1 Mouse Monoclonal Antibody [clone: TYRR1-1]
Proveedor: ProSci Inc.
34-001
PRSI34-001EA 773 EUR
PRSI34-001
Anti-TYRP1 Mouse Monoclonal Antibody [clone: TYRR1-1]
Anticuerpos
Tyrosinase-Related Protein-1/TYRP1/TRP1 is a melanocyte-specific gene product involved in melanin synthesis. While mouse Tyrp1 possesses dihydroxyindole carboxylic acid oxidase activity, the function in human melanocytes is less clear. In addition to its role in melanin synthesis, Tyrp1 is involved in stabilizing of tyrosinase protein and modulating its catalytic activity. Tyrp1 is also involved in maintenance of melanosome structure and affects melanocyte proliferation and melanocyte cell death. [Wiki].
Immunofluorescence: 3-6 µg/ml Immunohistochemistry (FFPE): 3-6 µg/ml for 30 min at RTMore Product Information