Anti-IRS1 Rabbit Polyclonal Antibody
Proveedor: ProSci Inc.
3521
PRSI3521EA 435 EUR
PRSI3521
Anti-IRS1 Rabbit Polyclonal Antibody
Anticuerpos
IRS-1 Antibody: Following tyrosine phosphorylation, the insulin receptor substrate 1 and 2 (IRS-1 and IRS-2) can form a protein scaffolding for the assembly of a host of Src homology 2 (SH2) domain-containing proteins. IRS-1 tyrosine phosphorylation can occur through the activity of several cytokine and growth factor receptors such as interleukin (IL)-4, IL-9, interferon-gamma, in addition to the insulin and insulin-like growth factor 1 receptors. The scaffolding provided by IRS-1 and IRS-2 is necessary for insulin signal transduction across the cell membrane. IRS-1 tyrosine phosphorylation,More Product Information