Anti-PIST Rabbit Polyclonal Antibody

Proveedor: ProSci Inc.
3633
PRSI3633EA 435 EUR
PRSI3633
Anti-PIST Rabbit Polyclonal Antibody
Anticuerpos
PIST Antibody: Autophagy, the process of bulk degradation of cellular proteins through an autophagosomic-lysosomal pathway is important for normal growth control and may be defective in tumor cells. It is involved in the preservation of cellular nutrients under starvation conditions as well as the normal turnover of cytosolic components and is negatively regulated by TOR (Target of rapamycin). PIST, a PDZ-containing protein, was discovered in a yeast two-hybrid system as a binding partner to Beclin-1, a Bcl-2-interacting protein homologous to the yeast autophagy gene apg6. Experiments withMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno PIST
Environmentally Preferable
Nombre del antígeno PDZ protein interacting specifically with TC10
Sinónimos de antígenos CFTR-associated ligand|Fused in glioblastoma|PDZ protein interacting specifically with TC10
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human,Mouse
Huésped Rabbit
Isotipo IgG
Western blot Yes
ELISA Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Inmunógeno PIST antibody was raised against a 17 amino acid synthetic peptide from near the center of human PIST.
Purification Affinity chromatography purified via peptide column
Storage buffer PBS containing 0.02% sodium azide
Tested applications E, WB, IHC, IF

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR