Human recombinant fumarase (from E. coli)
Proveedor: ProSci Inc.
91-204
PRSI91-204EA 884 EUR
PRSI91-204
Human recombinant fumarase (from E. coli)
Enzimas
Fumarase is an enzyme that catalyze the reversible hydration/dehydration of fumarate to S-malate and is involved in the tricarboxylic acid or Krebs cycle. Fumarase exists in both form, cytosolic formand N-terminal extend mitochondrial form. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension is the same form as in the cytoplasm. Fumarase is thought to act as a tumour suppressor, which deficiency can lead to progressive encephalopathy, cerebral atrophy and development delay.
This recombinant protein can be used for biological assays. For research use only.