Human recombinant alpha-Galactosidase A (from cells)
Proveedor: ProSci Inc.
91-323
PRSI91-323EA 734 EUR
PRSI91-323
Human recombinant alpha-Galactosidase A (from cells)
Enzimas
alpha-Galactosidase A is a homodimeric glycoprotein that belongs to the glycosyl hydrolase 27 family. It is a lysosomal enzyme and used as a long-term enzyme replacement therapy in patients with a confirmed diagnosis of Fabry disease. alpha-Galactosidase A can hydrolyze terminal alpha-galactosyl moieties from glycolipids and glycoproteins and catalyze the hydrolysis of melibiose into galactose and glucose. Defects alpha-Galactosidase A are the cause of Fabry disease (FD) which is a rare X-linked sphingolipidosis disease with glycolipid accumulates in many tissues. The disease consists of an inborn error of glycosphingolipid catabolism. FD patients show systemic accumulation of globotriaoslyceramide (Gb3) and related glycosphingolipids in the plasma and cellular lysosomes throughout the body. Patients may show ocular deposits, febrile episodes, and burning pain in the extremities. Death results from renal failure, cardiac or cerebral complications of hypertension or other vascular disease.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.