Human recombinant N-acetylgalactosaminyltransferase 7 (from cells)
Proveedor: ProSci Inc.
91-921
PRSI91-921EA 884 EUR
PRSI91-921
Human recombinant N-acetylgalactosaminyltransferase 7 (from cells)
Enzimas
N-acetylgalactosaminyltransferase 7(GALNT7) is expressed in uterus, retina, kidney, small intestine, omentum, stomach and CNS.It belongs to the glycosyltransferase 2 family and galNAc-T subfamily. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.