E. coli recombinant tryptophan synthase beta chain (from E. coli)
Proveedor: ProSci Inc.
92-623
PRSI92-623EA 654 EUR
PRSI92-623
E. coli recombinant tryptophan synthase beta chain (from E. coli)
Enzimas
Tryptophan synthase is an enzyme that catalyses the final two steps in the biosynthesis of tryptophan. It is commonly found in Eubacteria, Archaebacteria, Protista, Fungi, and Plantae, but is absent from animals such as humans. Tryptophan synthase typically exists as an alpha- beta beta - alpha complex.The alpha subunit is responsible for the aldol cleavage of indoleglycerol phosphate to indole and glyceraldehyde 3-phosphate: L-serine + 1-C-(indol-3-yl)glycerol 3-phosphate = L-tryptophan + D-glyceraldehyde 3-phosphate + H₂O.The beta subunits catalyse the irreversible condensation of indole and serine to form tryptophan in a pyridoxal phosphate (PLP) dependent reaction. Their assembly into a complex leads to structural changes in both subunits resulting in reciprocal activation.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.