Sistemas de evaporación al vacío RapidVap®
Proveedor: LABCONCO
7900001 7900003 7900011 7900013
531-0171EA 24100 EUR
531-0171 531-0172 531-0219 531-0220
Sistemas de evaporación al vacío RapidVap®
Evaporadores Evaporadores centrífugos
These systems use vacuum along with heat and vortex motion to help speed evaporation and provide an alternative to traditional centrifugal evaporation methods. The systems may be used with a broad range of aggressive chemicals. Heat is supplied by a dry block heating system that, unlike water baths, adds no potential source of contamination and requires no maintenance.
  • Alta velocidad de evaporación; el agua se evapora a una velocidad de 0,75 ml/minuto por tubo
  • Sistema de calentamiento de hasta +100 °C de bloque seco de 1000 vatios controlado por